ENA444724119 Enamine

4-(3-aminophenyl)-5-methyl-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD28348134
  • Cas Number : 142861-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202407
Availability details for
Quantity
  • Availability
  • Add to Cart