ENA444724123 Enamine

N-[(thiophen-2-yl)methyl]-1H-pyrazolo[3,4-d]pyrimidin-4-amine hydrochloride

  • MDL number MFCD28348138
  • Cas Number : 1803567-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202415
Availability details for
Quantity
  • Availability
  • Add to Cart