ENA444724130 Enamine

(2S)-2-{[1-(4-chlorophenyl)ethyl]amino}-3-methylbutan-1-ol hydrochloride

  • MDL number MFCD28348146
  • Cas Number : 1807920-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202553
Availability details for
Quantity
  • Availability
  • Add to Cart