ENA444724132 Enamine

1-(2-bromo-1,3-thiazol-5-yl)methanamine dihydrochloride

  • MDL number MFCD28348148
  • Cas Number : 1803600-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202561
Availability details for
Quantity
  • Availability
  • Add to Cart