ENA444724137 Enamine

4-(difluoromethyl)-5-methanesulfonyl-2H-1,2,3-triazole

  • MDL number MFCD28348153
  • Cas Number : 1803603-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202706
Availability details for
Quantity
  • Availability
  • Add to Cart