ENA444724140 Enamine

1-(4-tert-butylphenyl)-2-methylpropan-1-amine hydrochloride

  • MDL number MFCD28348156
  • Cas Number : 1803585-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202734
Availability details for
Quantity
  • Availability
  • Add to Cart