ENA444724147 Enamine

tert-butyl 2-(2-cyclopropyl-2-oxoethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD23318041
  • Cas Number : 1499728-69-6
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200573
Availability details for
Quantity
  • Availability
  • Add to Cart