ENA444724148 Enamine

3-[3-(thiophen-2-yl)-1,2,4-oxadiazol-5-yl]morpholine hydrochloride

  • MDL number MFCD28246478
  • Cas Number : 1803584-73-7
  • Molecular Weight : 273.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200574
Availability details for
Quantity
  • Availability
  • Add to Cart