ENA444724156 Enamine

N-(4-hydroxy-3-methylphenyl)pyridine-3-carboxamide

  • MDL number MFCD00723270
  • Cas Number : 1510533-24-0
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200583
Availability details for
Quantity
  • Availability
  • Add to Cart