ENA444724199 Enamine

(1R)-1-[2-(methylsulfanyl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD16293364
  • Molecular Weight : 203.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200700
Availability details for
Quantity
  • Availability
  • Add to Cart