ENA444724245 Enamine

2-hydroxy-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD16998747
  • Cas Number : 1243393-79-4
  • Molecular Weight : 203.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-199860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199860
Availability details for
Quantity
  • Availability
  • Add to Cart