ENA444724247 Enamine

2-[2-(methylamino)ethoxy]ethan-1-ol

  • MDL number MFCD12783671
  • Cas Number : 85475-01-0
  • Molecular Weight : 119.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200831

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-200831
pictograms GHS05,GHS07
hstatements H227,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart