ENA444724307 Enamine

1-(2-hydroxy-4,6-dimethylphenyl)ethan-1-one

  • MDL number MFCD00020128
  • Cas Number : 16108-50-2
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200989
Availability details for
Quantity
  • Availability
  • Add to Cart