ENA444724310 Enamine

methyl 2-hydroxy-2-[2-(trifluoromethyl)phenyl]acetate

  • MDL number MFCD16723802
  • Cas Number : 1250424-06-6
  • Molecular Weight : 234.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200997
Availability details for
Quantity
  • Availability
  • Add to Cart