ENA444724365 Enamine

2-{[(2-chlorophenyl)methyl](ethyl)amino}acetic acid hydrochloride

  • MDL number MFCD28348063
  • Cas Number : 1572282-39-3
  • Molecular Weight : 264.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200157
Availability details for
Quantity
  • Availability
  • Add to Cart