ENA444724528 Enamine

2-hydroxy-2-methyl-1-[4-(propan-2-yl)phenyl]propan-1-one

  • MDL number MFCD00036339
  • Cas Number : 69673-85-4
  • Molecular Weight : 206.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200266
Availability details for
Quantity
  • Availability
  • Add to Cart