ENA444724542 Enamine

2-[(3-aminophenyl)formamido]-N-methylacetamide

  • MDL number MFCD11638633
  • Cas Number : 1096890-80-0
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200273
Availability details for
Quantity
  • Availability
  • Add to Cart