ENA444724565 Enamine

5-chloro-3-methyl-1,2-thiazole-4-carbonitrile

  • MDL number MFCD28246450
  • Cas Number : 25069-00-5
  • Molecular Weight : 158.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200281
Availability details for
Quantity
  • Availability
  • Add to Cart