ENA444724671 Enamine

2-(benzyloxy)pyridin-4-amine

  • MDL number MFCD16084387
  • Cas Number : 1214900-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 97-98
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201919
Availability details for
Quantity
  • Availability
  • Add to Cart