ENA444724672 Enamine

4-bromo-2-(2,2,2-trifluoroethoxy)-1,3-thiazole

  • MDL number MFCD21869916
  • Cas Number : 1508037-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201920
Availability details for
Quantity
  • Availability
  • Add to Cart