ENA444724676 Enamine

4-bromo-1-[(4-methoxyphenyl)methyl]-1H-pyrazole

  • MDL number MFCD12824318
  • Cas Number : 1178371-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201932

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201932
Availability details for
Quantity
  • Availability
  • Add to Cart