ENA444724679 Enamine

2-(difluoromethoxy)-4-(trifluoromethyl)pyridine

  • MDL number MFCD13185859
  • Cas Number : 1214385-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201937
Availability details for
Quantity
  • Availability
  • Add to Cart