ENA444724680 Enamine

2-[(2-chlorothiophen-3-yl)oxy]acetic acid

  • MDL number MFCD20644639
  • Cas Number : 65449-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201938
Availability details for
Quantity
  • Availability
  • Add to Cart