ENA444724698 Enamine

ethyl 2-bromo-6-(bromomethyl)benzoate

  • MDL number MFCD18205384
  • Cas Number : 1261851-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201977
Availability details for
Quantity
  • Availability
  • Add to Cart