ENA444724702 Enamine

3-methyl-4-(trifluoromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD18399870
  • Cas Number : 1261753-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202004
Availability details for
Quantity
  • Availability
  • Add to Cart