ENA444724704 Enamine

2-(3-amino-4-methylphenyl)-1lambda6,2-thiazolidine-1,1-dione

  • MDL number MFCD08443630
  • Cas Number : 889940-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202045
Availability details for
Quantity
  • Availability
  • Add to Cart