ENA444724706 Enamine

2-(bromomethyl)-4-methylpentanenitrile

  • MDL number MFCD18294599
  • Cas Number : 1273849-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202047
Availability details for
Quantity
  • Availability
  • Add to Cart