ENA444724712 Enamine

1-(1H-1,3-benzodiazol-2-yl)-2-methylpropan-2-amine

  • MDL number MFCD20386483
  • Cas Number : 39650-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202067
Availability details for
Quantity
  • Availability
  • Add to Cart