ENA444724714 Enamine

(1R)-1-(3-bromophenyl)-2-chloroethan-1-ol

  • MDL number MFCD24346571
  • Cas Number : 1567881-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202071
Availability details for
Quantity
  • Availability
  • Add to Cart