ENA444724721 Enamine

2-chloro-N-(cyclopropylmethyl)pyridin-4-amine

  • MDL number MFCD23924449
  • Cas Number : 1549879-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202082
Availability details for
Quantity
  • Availability
  • Add to Cart