ENA444724735 Enamine

methyl 4-amino-1,1-dioxo-1lambda6-thiane-4-carboxylate hydrochloride

  • MDL number MFCD22375544
  • Cas Number : 1363992-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202143
Availability details for
Quantity
  • Availability
  • Add to Cart