ENA444724736 Enamine

1-(pyridin-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD17277883
  • Cas Number : 896135-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202144

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202144
Availability details for
Quantity
  • Availability
  • Add to Cart