ENA444724742 Enamine

N-methyl-2-(methylamino)-N-(1-phenylethyl)acetamide hydrochloride

  • MDL number MFCD28246601
  • Cas Number : 1605816-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202153
Availability details for
Quantity
  • Availability
  • Add to Cart