ENA444724751 Enamine

(5-fluoro-2-nitrophenyl)methanesulfonyl chloride

  • MDL number MFCD12783405
  • Cas Number : 1184150-93-3
  • Molecular Weight : 253.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200352
Availability details for
Quantity
  • Availability
  • Add to Cart