ENA444724755 Enamine

2-amino-3-cyclopentylpropan-1-ol hydrochloride

  • MDL number MFCD08455587
  • Cas Number : 1380005-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202200
Availability details for
Quantity
  • Availability
  • Add to Cart