ENA444724758 Enamine

3-(2,2-dimethylpropyl)-6-ethyl-[1,2]oxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD12458048
  • Cas Number : 1096901-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202204
Availability details for
Quantity
  • Availability
  • Add to Cart