ENA444724763 Enamine

propan-2-yl 3,4-dihydroxybenzoate

  • MDL number MFCD24390050
  • Cas Number : 105603-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202212

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202212
Availability details for
Quantity
  • Availability
  • Add to Cart