ENA444724766 Enamine

7-ethyl-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD15525621
  • Cas Number : 73425-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202215
Availability details for
Quantity
  • Availability
  • Add to Cart