ENA444724773 Enamine

3-(oxolan-3-yl)prop-2-ynoic acid

  • MDL number MFCD20347401
  • Cas Number : 1343209-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202225
Availability details for
Quantity
  • Availability
  • Add to Cart