ENA444724776 Enamine

ethyl 5-chloropyrazolo[1,5-a]pyridine-2-carboxylate

  • MDL number MFCD20265008
  • Cas Number : 1691046-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202230
Availability details for
Quantity
  • Availability
  • Add to Cart