ENA444724777 Enamine

5-chloropyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD20258706
  • Cas Number : 1545047-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202231
Availability details for
Quantity
  • Availability
  • Add to Cart