ENA444724782 Enamine

N-(3-aminopropyl)-3-phenylprop-2-enamide hydrochloride

  • Cas Number : 1607781-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202244
Availability details for
Quantity
  • Availability
  • Add to Cart