ENA444724801 Enamine

8-bromoquinolin-3-ol

  • MDL number MFCD18413588
  • Cas Number : 1261768-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202327
Availability details for
Quantity
  • Availability
  • Add to Cart