ENA444724804 Enamine

2-(2-bromoethoxy)propanenitrile

  • MDL number MFCD20625063
  • Cas Number : 854679-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202330
Availability details for
Quantity
  • Availability
  • Add to Cart