ENA444724806 Enamine

(1-cyclopropylethyl)[(2,4-dimethoxyphenyl)methyl]amine

  • MDL number MFCD12427882
  • Cas Number : 1155988-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202332
Availability details for
Quantity
  • Availability
  • Add to Cart