ENA444724809 Enamine

2-amino-N-(3,4-dihydro-2H-1-benzopyran-4-yl)acetamide hydrochloride

  • MDL number MFCD28348125
  • Cas Number : 1579143-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202338
Availability details for
Quantity
  • Availability
  • Add to Cart