ENA444724821 Enamine

(2R)-2-(dimethylamino)propan-1-ol

  • MDL number MFCD26906333
  • Cas Number : 55197-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202357
Availability details for
Quantity
  • Availability
  • Add to Cart