ENA444724824 Enamine

4-acetyl-3-hydroxybenzoic acid

  • MDL number MFCD00854034
  • Cas Number : 102297-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202364
Availability details for
Quantity
  • Availability
  • Add to Cart