ENA444724825 Enamine

5-chloro-3-(1-methoxyethyl)-1,2,4-thiadiazole

  • MDL number MFCD21874771
  • Cas Number : 1506875-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202365
Availability details for
Quantity
  • Availability
  • Add to Cart