ENA444724826 Enamine

3-amino-3-ethylpentan-1-ol

  • MDL number MFCD21184469
  • Cas Number : 1487552-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202366
Availability details for
Quantity
  • Availability
  • Add to Cart